missing translation for 'onlineSavingsMsg'
Learn More

Notch-4 Intracellular Domain Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3847

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438404

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Notch-4 Intracellular Domain Polyclonal antibody AF3847 Specifically detects Notch-4 Intracellular Domain in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Notch-4
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q99466
INT3, neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3, Notch4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Notch-4 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-1 Intracellular Domain and rhNotch-2 Intracellular Domain is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NOTCH4
E. coli-derived recombinant human Notch-4 Intracellular Domain Gly1778-Lys2003 Accession # Q99466
Immunogen affinity purified
RUO
Primary
4855
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only