Learn More
Notch-3 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1559
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487004
  • £382.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Notch-3 Polyclonal antibody AF1559 Specifically detects Notch-3 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications
Specifications
Notch-3
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
recombinant human Notch-3 extracellular domain, Ala40-Glu467, Accession No. Q9UM47
100μg
Primary
4854
Unconjugated
Q9UM47
Sheep