missing translation for 'onlineSavingsMsg'
Learn More

Notch-2 Intracellular Domain Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3735

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458334

  • £314.17 / 100µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Notch-2 Intracellular Domain Polyclonal antibody AF3735 Specifically detects Notch-2 Intracellular Domain in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Notch-2
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q04721
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Notch-2 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Notch-3 (aa 2195-2321), rhNotch-6 (aa 2251-2556) and rhNotch-4 (aa 1778-2003) is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NOTCH2
E. coli-derived recombinant human Notch-2 Asp2063-Pro2413 Accession # Q04721
Antigen Affinity-purified
RUO
Primary
4853
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only