missing translation for 'onlineSavingsMsg'
Learn More

Norrin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3014

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447924

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 28-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Norrin Polyclonal antibody AF3014 Specifically detects Norrin in Human samples. It is validated for Blocking (Receptor-ligand), Immunohistochemistry, Western Blotting
Specifications

Specifications

Norrin/NDP
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL
Q00604
EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, NDP, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Norrin in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Norrin is observed.
Human
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NDP
E. coli-derived recombinant human Norrin Lys25-Ser133 Accession # Q00604
Immunogen affinity purified
RUO
Primary
4693
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only