missing translation for 'onlineSavingsMsg'
Learn More

Nogo Receptor/NgR Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1208

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436814

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nogo Receptor/NgR Polyclonal antibody AF1208 Specifically detects Nogo Receptor/NgR in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

Nogo Receptor/NgR
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q9BZR6
NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Nogo Receptor (NgR) in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant mouse NgR is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RTN4R
recombinant human NgR Cys27-Ser447 Accession # Q9BZR6
Antigen Affinity-purified
RUO
Primary
65078
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only