Learn More
NKX2.5 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2444
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467564
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
NKX2.5 Polyclonal antibody AF2444 Specifically detects NKX2.5 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
NKX2.5
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
P52952
cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2-5, NKX2E, NKX2EFLJ97197, NKX4-1, tinman paralog
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human NKX2.5 in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NKX2-5
E. coli-derived recombinant human NKX2.5 Gln24-Arg137 Accession # P52952
Immunogen affinity purified
RUO
Primary
1482
Reconstitute at 0.2 mg/mL in sterile PBS.