missing translation for 'onlineSavingsMsg'
Learn More

NF-M Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3029

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437934

  • £298.33 / 100µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NF-M Polyclonal antibody AF3029 Specifically detects NF-M in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

NF-M
LYOPH
Western Blot 1-2 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
P07197
NEF3, NEF3neurofilament medium polypeptide, NEFM, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NFM, NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human NF-M in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) NF-H and rhNF-L is observed.
Human
Western Blot, Immunohistochemistry, KnockDown
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NEFM
E. coli-derived recombinant human NF-M Ser2-Ser358 Accession # P07197
Antigen Affinity-purified
RUO
Primary
4741
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only