missing translation for 'onlineSavingsMsg'
Learn More

Neprilysin-2/MMEL1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2340

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457494

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Neprilysin-2/MMEL1 Polyclonal antibody AF2340 Specifically detects Neprilysin-2/MMEL1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Neprilysin-2/MMEL1
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready
AAL08942
EC 3.4.24, EC 3.4.24.11, MELL1, membrane metallo-endopeptidase-like 1, Membrane metallo-endopeptidase-like 2MGC119455, MGC119454, MMEL1, MMEL2, NEP2, NEP2(m), NEPIIMGC119456, Neprilysin II, neprilysin-2, Neprilysin2, NL2, NL2NL1, SEP, soluble secreted endopeptidase, zinc metallopeptidase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Neprilysin-2/MMEL1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Neprilysin is observed and less than 1% cross-reactivity with rhECE-1 and rhECE-2 is observed.
Human
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MMEL1
Mouse myeloma cell line NS0-derived recombinant human Neprilysin-2/MMEL1 Gly69-Trp770 Accession # AAL08942
Immunogen affinity purified
RUO
Primary
79258
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only