missing translation for 'onlineSavingsMsg'
Learn More

Wnt-2b Goat anti-Human, Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3900

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468434

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Wnt-2b Polyclonal antibody AF3900 Specifically detects Wnt-2b in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Wnt-2b
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
O70283
Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, Wnt-13, WNT13XWNT2, Xenopus, homolog of, Wnt2b, XWNT2, XWNT2wingless-type MMTV integration site family, member 13
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects recombinant human and recombinant mouse Wnt-2b in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Wnt-2 is observed and less than 5% cross-reactivity with rhWnt-7a, -9a, recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, and -16 is observed.
Human, Mouse
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
WNT2B
E. coli-derived recombinant mouse Wnt-2b Leu64-Glu104 and Arg251-Ala312 Accession # O70283
Immunogen affinity purified
RUO
Primary
7482
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only