missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6B Goat anti-Human, Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2094

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447344

  • £433.65 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6B Polyclonal antibody AF2094 Specifically detects Semaphorin 6B in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Semaphorin 6B
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q9H3T3
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, Sema6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, SEMAZ, SEM-SEMA-Y, UNQ1907/PRO4353
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse Semaphorin 6B in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed, 5% cross-reactivity with rhSemaphorin 6D is observed and less than 1% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, rhSemaphorin 6A, rhSemaphorin 7A, recombinant mouse (rm) Semaphorin 3C, rmSemaphorin 3E, and rmSemaphorin 3F is observed.
Human, Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SEMA6B
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6B Leu26-Ser603 Accession # Q9H3T3
Immunogen affinity purified
RUO
Primary
10501
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only