missing translation for 'onlineSavingsMsg'
Learn More

MKK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1604

Code : Z1

Additional Details : Weight : 0.00010kg

Product Code. 13487034

  • £302.29 / 50µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

MKK6 Polyclonal antibody AF1604 Specifically detects MKK6 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications

Specifications

MKK6/MEK6
LYOPH
Western Blot 0.05 ug/mL
AAC50388
MAP kinase kinase 6, MAP2K6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3, SAPKK3EC 2.7.12.2
Rabbit
IgG
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human, mouse, and rat MKK6 in Western blots.
Human, Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MAP2K6
E. coli-derived recombinant human MKK6, Gly6-Asp334, Accession # AAC50388
Immunogen affinity purified
RUO
Primary
5608
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos