missing translation for 'onlineSavingsMsg'
Learn More

Lipin 2 Goat anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3756

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458344

  • £314.17 / 100µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Lipin 2 Polyclonal antibody AF3756 Specifically detects Lipin 2 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications

Specifications

Lipin 2
LYOPH
Western Blot 0.1 ug/mL
Q92539
EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, LPIN2, phosphatidate phosphatase LPIN2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects Lipin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Lipin 1 and rhLipin 3 is observed.
Human, Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LPIN2
E. coli-derived recombinant human Lipin 2 Ser439-Lys563 Accession # Q92539
Immunogen affinity purified
RUO
Primary
9663
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only