missing translation for 'onlineSavingsMsg'
Learn More

EG-VEGF/PK1 Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487344

  • £382.00 / 100µg


Estimated Shipment
07-03-2018
Add to basket

Disclaimers:

For Research Use Only


Description

Description

EG-VEGF/PK1 Polyclonal antibody AF2100 Specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications

Specifications

EG-VEGF/PK1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant rat EG-VEGF/PK1, Ala19-Phe105, Accession # Q8R414
100μg
Primary
192205
Rat
Unconjugated
Q8R414
Sheep
IgG
RUO
Polyclonal
Detects human, mouse, and rat EG-VEGF/PK1 in direct ELISAs and Western blots. In direct ELISAs, 100% cross reactivity with recombinant human EG-VEGF and recombinant mouse EG-VEGF is observed
Videos