missing translation for 'onlineSavingsMsg'
Learn More

Akt2 Rabbit anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2315

Code : Z1

Additional Details : Weight : 0.00010kg

Product Code. 13487474

  • £302.29 / 50µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Akt2 Polyclonal antibody AF2315 Specifically detects Akt2 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications

Specifications

AKT2
LYOPH
Western Blot 0.2 ug/mL, Simple Western 2.5 ug/mL
P31751
EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2
Rabbit
IgG
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human, mouse, and rat Akt2 in Western blots. In Western blots, this antibody also detects recombinant human (rh) Akt2 but not rhAkt1 or rhAkt3.
Human, Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AKT2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant recombinant human Akt2, Accession # P31751
Antigen Affinity-purified
RUO
Primary
208
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos