Learn More
Akt1 Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF17751
Code : Z1
Additional Details : Weight : 0.00010kg
Product Code. 13487144
  • £382.00 / 100µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Akt1 Polyclonal antibody AF17751 Specifically detects Akt1 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
Akt1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant human Akt1, Met1-Glu149, Accession No. P31749
100μg
Primary
25233
Unconjugated
P31749
Sheep
IgG
RUO
Polyclonal
HumanþMurineþRat
Videos