missing translation for 'onlineSavingsMsg'
Learn More

Nectin-1 Goat anti-Human, Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2880

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447844

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nectin-1 Polyclonal antibody AF2880 Specifically detects Nectin-1 in Human, Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Nectin-1/PVRL1
LYOPH
Western Blot 0.1 ug/mL
Q15223
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin1, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1, PRR1MGC16207, PVRL1, PVRR1, PVRR1nectin-1, SK-12
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse Nectin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 6% cross-reactivity with recombinant human (rh) Nectin-2, -3, and -4 is observed.
Human, Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PVRL1
Mouse myeloma cell line NS0-derived recombinant human Nectin-1 Gln31-Thr334 Accession # Q15223
Immunogen affinity purified
RUO
Primary
5818
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only