Learn More
Lefty Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF746
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430125
  • £454.23 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Lefty Polyclonal antibody AF746 Specifically detects Lefty in Human, Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Lefty
LYOPH
Western Blot 0.1 ug/mL
Q64280
LEFTY2
Mouse myeloma cell line NS0-derived recombinant mouse Lefty-1 Leu22-Arg135, His6, Leu136-Pro368 Accession # Q64280
Immunogen affinity purified
RUO
Primary
7044
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse Lefty in direct ELISAs and Western blots.
Human, Mouse