missing translation for 'onlineSavingsMsg'
Learn More

BACE-1 Ectodomain Goat anti-Human, Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF931

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440335

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BACE-1 Ectodomain Polyclonal antibody AF931 Specifically detects BACE-1 Ectodomain in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

BACE-1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P56817
APP beta-secretase, ASP2, Aspartyl protease 2, BACE1, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse BACE-1 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) ADAM8, recombinant mouse ADAM9, rhADAM15, and rhTACE is observed.
Human, Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BACE1
Mouse myeloma cell line NS0-derived recombinant human BACE-1 Ectodomain Thr22-Tyr460 Accession # P56817
Immunogen affinity purified
RUO
Primary
23621
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only