missing translation for 'onlineSavingsMsg'
Learn More

MIS/AMH Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1737

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437124

  • £314.17 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MIS/AMH Polyclonal antibody AF1737 Specifically detects MIS/AMH in Human samples. It is validated for Western Blotting
Specifications

Specifications

MIS/AMH
LYOPH
Western Blot 0.1 ug/mL
Q6GTN3
AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MIS/AMH in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant rat MIS and recombinant mouse MIS is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
AMH
E. coli-derived recombinant human MIS/AMH (R&D Systems, Catalog # 1737-MS) Ala453-Arg560 Accession # Q6GTN3
Antigen Affinity-purified
RUO
Primary
268
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only