Learn More
Mindin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2609
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13437674
  • £449.92 / 100µg
Estimated Shipment
11-09-2020
Description
Description
Mindin Polyclonal antibody AF2609 Specifically detects Mindin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Mindin/Spondin-2
LYOPH
Western Blot 0.1 ug/mL
Q9BUD6
Differentially expressed in cancerous and non-cancerous lung cells 1, DIL1, DIL-1, DIL1FLJ34460, DKFZp686G21139, FLJ16313, FLJ22401, MINDIN, M-SPONDIN, RG-1, SPON2, Spondin 2, spondin 2, extracellular matrix protein, spondin-2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Mindin in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SPON2
Mouse myeloma cell line NS0-derived recombinant human Mindin Glu32-Val331 Accession # Q9BUD6
Immunogen affinity purified
RUO
Primary
10417
Reconstitute at 0.2 mg/mL in sterile PBS.