missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICL/CLEC12A Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: RD Systems (Bio-Techne) AF2946
Code : Z1
Additional Details : Weight : 0.00010kg
Description
MICL/CLEC12A Polyclonal antibody AF2946 Specifically detects MICL/CLEC12A in Human samples. It is validated for Immunocytochemistry, Western BlottingSpecifications
MICL/CLEC12A | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
NP_612210 | |
CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
Goat | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human MICL/CLEC12A in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse MICL is observed. | |
Human |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
CLEC12A | |
Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A isoform a Thr67-Ala265 Accession # NP_612210 | |
Immunogen affinity purified | |
RUO | |
Primary | |
160364 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
For Research Use Only