missing translation for 'onlineSavingsMsg'
Learn More

MFRP C-Terminus Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1915

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467224

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MFRP C-Terminus Polyclonal antibody AF1915 Specifically detects MFRP C-Terminus in Human samples. It is validated for Western Blotting
Specifications

Specifications

MFRP
LYOPH
Western Blot 0.1 ug/mL
Q9BY79
MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MFRP C-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-4, recombinant human Frizzled-5, rmFrizzled-6, rmFrizzled-7, rmFrizzled-8, and rmFrizzled-9 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MFRP
Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579 Accession # Q9BY79
Immunogen affinity purified
RUO
Primary
83552
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only