missing translation for 'onlineSavingsMsg'
Learn More

Mesothelin Propeptide/MPF Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3448

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448174

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mesothelin Propeptide/MPF Polyclonal antibody AF3448 Specifically detects Mesothelin Propeptide/MPF in Human samples. It is validated for Western Blotting
Specifications

Specifications

Mesothelin
LYOPH
Western Blot 0.1 ug/mL
Q13421
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Mesothelin Propeptide/MPF in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant human Mesothelin is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MSLN
Mouse myeloma cell line NS0-derived recombinant human Mesothelin Propeptide/MPF isoform 1 Arg35-Leu289 Accession # Q13421
Immunogen affinity purified
RUO
Primary
10232
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only