missing translation for 'onlineSavingsMsg'
Learn More

Meprin beta Subunit/MEP1B Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2895

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437854

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Meprin beta Subunit/MEP1B Polyclonal antibody AF2895 Specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Meprin beta Subunit/MEP1B
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
NP_005916
EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Meprin beta Subunit/MEP1B in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse MEP1B is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MEP1B
Mouse myeloma cell line NS0-derived recombinant human Meprin beta Subunit/MEP1B Leu21-Ser593 Accession # NP_005916
Immunogen affinity purified
RUO
Primary
4225
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only