missing translation for 'onlineSavingsMsg'
Learn More

Meprin alpha Subunit/MEP1A Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3220

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488044

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Meprin alpha Subunit/MEP1A Polyclonal antibody AF3220 Specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Meprin alpha Subunit/MEP1A
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
AAA21338
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse MEP1A and recombinant human MEP1B is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MEP1A
Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A, Val22-Gln601, Accession # AAA21338
Immunogen affinity purified
RUO
Primary
4224
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only