Learn More
Melusin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2740
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457754
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Melusin Polyclonal antibody AF2740 Specifically detects Melusin in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Melusin/ITGB1BP2
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q9UKP3
CHORDC3, integrin beta 1 binding protein (melusin) 2, integrin beta-1-binding protein 2, ITGB1BP, ITGB1BP2, Melusin, MGC119214
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Melusin in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ITGB1BP2
E. coli-derived recombinant human Melusin Ser2-Glu347 Accession # Q9UKP3
Antigen Affinity-purified
RUO
Primary
26548
Reconstitute at 0.2 mg/mL in sterile PBS.