missing translation for 'onlineSavingsMsg'
Learn More

MDGA2 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5184

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449134

  • £344.84 / 100µg

Estimated Shipment date
from Supplier 08-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MDGA2 Polyclonal antibody AF5184 Specifically detects MDGA2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

MDGA2
LYOPH
Western Blot 0.1 ug/mL
Q7Z553
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MDGA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human MDGA1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MDGA2
Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp931 Accession # Q7Z553
Antigen Affinity-purified
RUO
Primary
161357
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only