missing translation for 'onlineSavingsMsg'
Learn More

MD-2 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1787

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447154

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 28-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MD-2 Polyclonal antibody AF1787 Specifically detects MD-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

MD-2
LYOPH
Western Blot 0.1 ug/mL
NP_001123946
ESOP1, ESOP-1, Ly-96, LY96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MD-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human MD-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LY96
E. coli-derived recombinant human MD-2 Glu17-Asn160 Accession # NP_001123946
Immunogen affinity purified
RUO
Primary
23643
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only