missing translation for 'onlineSavingsMsg'
Learn More

Mcl-1 Sheep anti-Human, Polyclonal, R&D Systems™

Brand:  RD Systems (Bio-Techne) AF8281

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460255

  • £314.17 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Mcl-1 Polyclonal antibody AF8281 Specifically detects Mcl-1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Mcl-1
LYOPH
Western Blot 0.5 ug/mL
Q07820
bcl2-L-3, BCL2L3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Mcl-1 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MCL1
E. coli-derived recombinant human Mcl-1, Val147-Gly219, Accession # Q07820
Antigen Affinity-purified
RUO
Primary
4170
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos