missing translation for 'onlineSavingsMsg'
Learn More

MCEMP1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4994

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13489024

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

MCEMP1 Polyclonal antibody AF4994 Specifically detects MCEMP1 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

MCEMP1
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q8IX19
C19orf59, chromosome 19 open reading frame 59, mast cell-expressed membrane protein 1, MCEMP1, MGC132456
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human MCEMP1 in direct ELISAs and Western blots.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
C19ORF59
Mouse myeloma cell line NS0-derived recombinant human MCEMP1, Val106-Gln187, Accession # Q8IX19
Antigen Affinity-purified
RUO
Primary
199675
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only