missing translation for 'onlineSavingsMsg'
Learn More

MBL Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2307

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437474

  • £449.92 / 100µg

Estimated Shipment date
from Supplier 10-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MBL Polyclonal antibody AF2307 Specifically detects MBL in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

MBL
LYOPH
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
AAH96182
COLEC1, COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBL2, MBL2D, MBLmannan-binding lectin, MBP, MBP1, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human MBL in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant mouse (rm) MBL and rmMBL-2 is observed.
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MBL2
Mouse myeloma cell line NS0-derived recombinant human MBL Glu21-Ile248 Accession # AAH96182
Immunogen affinity purified
RUO
Primary
4153
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only