missing translation for 'onlineSavingsMsg'
Learn More

Matrilin-3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3017

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467924

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Matrilin-3 Polyclonal antibody AF3017 Specifically detects Matrilin-3 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

NDUFC2
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Affinity Purified
NDUFC2
Mouse myeloma cell line NS0-derived recombinant human Matrilin-3
Antigen Affinity-purified
RUO
Primary
4718
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
B14.5b, CI-B14.5b, complex I subunit B14.5b, Complex I-B14.5b, EDM5, HLC-1NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2 (14.5kD, B14.5b), HOA, Human lung cancer oncogene 1 protein, Matrilin3, NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa, NADH dehydrogenase [ubiquinone] 1 subunit C2, NADHDH2, NADH-ubiquinone oxidoreductase subunit B14.5b, NDUFC2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Matrilin-3 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Matrilin-3 is observed and less than 1% cross-reactivity with recombinant human (rh) Matrilin-2 and rhMatrilin-4 is observed.
Human
Videos
Documents

For Research Use Only