missing translation for 'onlineSavingsMsg'
Learn More

Matrilin-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3044

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447944

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Matrilin-2 Polyclonal antibody AF3044 Specifically detects Matrilin-2 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Matrilin-2
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
AAH10444
MATN2, matrilin 2, matrilin-2, Matrilin2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Matrilin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse (rm) Matrilin-2 is observed and approximately 15% cross-reactivity with rmMatrilin-3 and recombinant human Matrilin-4 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MATN2
Mouse myeloma cell line NS0-derived recombinant human Matrilin-2 short isoform Arg24-Arg937 Accession # AAH10444
Antigen Affinity-purified
RUO
Primary
4147
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only