missing translation for 'onlineSavingsMsg'
Learn More

MAGP-2/MFAP5 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4914

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488974

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

MAGP-2/MFAP5 Polyclonal antibody AF4914 Specifically detects MAGP-2/MFAP5 in Human samples. It is validated for Western Blotting
Specifications

Specifications

MAGP-2/MFAP5
LYOPH
Western Blot 0.1 ug/mL
Q13361
MAGP2, MAGP-2, MAGP2MFAP-5, MFAP5, Microfibril-associated glycoprotein 2, microfibrillar associated protein 5, microfibrillar-associated protein 5, MP25, MP25microfibril-associated glycoprotein-2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human MAGP-2/MFAP5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human MAGP-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MFAP5
Mouse myeloma cell line NS0-derived recombinant human MAGP-2/MFAP5, Ile22-Leu173, Accession # Q13361
Immunogen affinity purified
RUO
Primary
8076
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only