missing translation for 'onlineSavingsMsg'
Learn More

M-CSF R Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF329

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488084

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

M-CSF R Polyclonal antibody AF329 Specifically detects M-CSF R in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

M-CSF R/CD115
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
CAA27300
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human M-CSF R/CD115 in direct ELISAs and Western blots.
Human
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CSF1R
Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115, Ile20-Glu512, Accession # CAA27300
Antigen Affinity-purified
RUO
Primary
1436
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only