missing translation for 'onlineSavingsMsg'
Learn More

Lymphotoxin beta R/TNFRSF3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF629

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459684

  • £389.41 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Lymphotoxin beta R/TNFRSF3 Polyclonal antibody AF629 Specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Flow Cytometry, Inhibition (Cell Growth), Western Blotting
Specifications

Specifications

Lymphotoxin beta R/TNFRSF3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Inhibition of Cell Growth, CyTOF-ready
P36941
CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Lymphotoxin beta R/TNFRSF3 in direct ELISAs and Western blots.
Human
Western Blot, Flow Cytometry, Inhibition Assays, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LTBR
Mouse myeloma cell line NS0-derived recombinant human Lymphotoxin beta R/TNFRSF3, Ser28-Met227, Accession # P36941
Immunogen affinity purified
RUO
Primary
4055
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only