missing translation for 'onlineSavingsMsg'
Learn More

LINGO-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3679

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488304

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

LINGO-2 Polyclonal antibody AF3679 Specifically detects LINGO-2 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

LINGO-2
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q7L985
LERN3, LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LINGO2, LRRN6C, LRRN6Cleucine rich repeat neuronal 6C
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human LINGO-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human LINGO-1 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LINGO2
Mouse myeloma cell line NS0-derived recombinant human LINGO-2, Cys28-Leu542, Accession # Q7L985
Immunogen affinity purified
RUO
Primary
158038
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only