Learn More
LIMPII/SR-B2 Lumenal Loop Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1966
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487254
  • £382.00 / 100µg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
LIMPII/SR-B2 Lumenal Loop Polyclonal antibody AF1966 Specifically detects LIMPII/SR-B2 Lumenal Loop in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
LIMPII/SR-B2 Lumenal Loop
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human LIMPII lumenal loop, Arg27-Thr432, Accession No. Q14108
100μg
Primary
950
Unconjugated
Q14108
Goat