Learn More
LECT2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF722
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460035
  • £298.33 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
LECT2 Polyclonal antibody AF722 Specifically detects LECT2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
LECT2
LYOPH
Western Blot 0.1 ug/mL
O14960
chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human LECT2 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LECT2
E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960
Immunogen affinity purified
RUO
Primary
3950
Reconstitute at 0.2 mg/mL in sterile PBS.