Learn More
Latexin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3246
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488054
  • £347.17 / 100µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Latexin Polyclonal antibody AF3246 Specifically detects Latexin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Latexin
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant human Latexin, Glu2-Glu222, Accession No. AAH05346
100μg
Primary
56925
Unconjugated
AAH05346
Goat