Learn More
LAIR1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2664
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487704
  • £382.00 / 100µg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
LAIR1 Polyclonal antibody AF2664 Specifically detects LAIR1 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
LAIR1
Unconjugated
Q6GTX8
Goat
IgG
RUO
Polyclonal
Human
CyTOFþFlow CytometryþWestern Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1, Gln22-His163, Accession No. Q6GTX8
100μg
Primary
3903