missing translation for 'onlineSavingsMsg'
Learn More

KLF6 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3499

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448204

  • £349.86 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

KLF6 Polyclonal antibody AF3499 Specifically detects KLF6 in Human samples. It is validated for Western Blotting
Specifications

Specifications

KLF6
LYOPH
Western Blot 0.1 ug/mL
Q99612
BCD1, BCD1Zf9, B-cell-derived protein 1, COPEB, COPEBCBA1, core promoter element binding protein, Core promoter element-binding protein, CPBP, CPBPZF9, DKFZp686N0199, EC 2.1.1.43, EC 6.1.1.15, GBF, GC-rich binding factor, GC-rich sites-binding factor GBF, Krueppel-like factor 6, Kruppel-like factor 6, Kruppel-like zinc finger protein Zf9, PAC1, Proto-oncogene BCD1, protooncogene B-cell derived 1, ST12, suppression of tumorigenicity 12 (prostate), Suppressor of tumorigenicity 12 protein, Transcription factor Zf9
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human KLF6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows less than 5% cross-reactivity with rhKLF4, rhKLF5 and rmKLF15.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLF6
E. coli-derived recombinant human KLF6 Met1-Ser109 Accession # Q99612
Immunogen affinity purified
RUO
Primary
1316
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only