missing translation for 'onlineSavingsMsg'
Learn More

Kirrel2/NEPH3 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2564

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437644

  • £334.48 / 100µg

Estimated Shipment
11-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kirrel2/NEPH3 Polyclonal antibody AF2564 Specifically detects Kirrel2/NEPH3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Kirrel2/NEPH3
LYOPH
Western Blot 0.1 ug/mL
Q6UWL6
DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kirrel2/NEPH3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Kirrel2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KIRREL2
Mouse myeloma cell line NS0-derived recombinant human Kirrel2/NEPH3 Gly21-Arg503 Accession # Q6UWL6
Antigen Affinity-purified
RUO
Primary
84063
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only