missing translation for 'onlineSavingsMsg'
Learn More

Kell Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1914

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457224

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kell Polyclonal antibody AF1914 Specifically detects Kell in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Kell
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready
Affinity Purified
KEL
Mouse myeloma cell line NS0-derived recombinant human Kell aa 68-732
Antigen Affinity-purified
RUO
Primary
3792
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kell in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse Kell is observed.
Human
Videos
Documents

For Research Use Only