missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 8/Neuropsin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437294

  • £449.92 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 8/Neuropsin Polyclonal antibody AF2025 Specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Neutralization, Western Blotting
Specifications

Specifications

Kallikrein 8/Neuropsin
LYOPH
Western Blot 0.1 ug/mL, Neutralization 8 ug/mL
O60259
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kallikrein 8/Neuropsin in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) KLK1, -6, -7, -11, -13, -14 and rhKLK15 is observed.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLK8
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8 Gln29-Gly260 Accession # O60259
Antigen Affinity-purified
RUO
Primary
11202
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only