missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 5 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1108

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436744

  • £371.48 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 5 Polyclonal antibody AF1108 Specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Kallikrein 5
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Q9Y337
EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLK5, KLKL2, KLK-L2, KLK-L2EC 3.4.21.-, SCTE, SCTEkallikrein-5, Stratum corneum tryptic enzyme
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kallikrein 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) KLK8 and rhKLK13 is observed and less than 1% cross-reactivity with rhKLK1, -3, -4, -7, -9, -10, -11, and -12 is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLK5
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 5 Ile67-Ser293 Accession # Q9Y337
Immunogen affinity purified
RUO
Primary
25818
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only