missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 14 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2626

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437684

  • £314.17 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 14 Polyclonal antibody AF2626 Specifically detects Kallikrein 14 in Human samples. It is validated for ELISA Capture, Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Kallikrein 14
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
AAD50773
EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kallikrein 14 in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 7, 8, 11, 12, and 13 is observed.
Human
Western Blot, Immunoprecipitation, Neutralization, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLK14
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248, predicted Accession # AAD50773
Immunogen affinity purified
RUO
Primary
43847
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only