missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 12 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3095

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467974

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 12 Polyclonal antibody AF3095 Specifically detects Kallikrein 12 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications

Specifications

Kallikrein 12
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Q9UKR0
DKFZp686H1078, EC 3.4.21, EC 3.4.21.-, kallikrein 12, Kallikrein-like protein 5, kallikrein-related peptidase 12, KLK12, KLK-L5, KLKL5kallikrein-12, KLK-L5MGC42603
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kallikrein 12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Kallikrein 8, rhKallikrein 10, rhKallikrein 11, and rhKallikrein 15 is observed.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLK12
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0
Antigen Affinity-purified
RUO
Primary
43849
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only