missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 10 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2007

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447284

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 10 Polyclonal antibody AF2007 Specifically detects Kallikrein 10 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Kallikrein 10
LYOPH
Western Blot 0.1 ug/mL
O43240
breast normal epithelial cell associated serine protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.74, Kallikrein 10, kallikrein-related peptidase 10, KLK10, NES1, NES1kallikrein-10, Normal epithelial cell-specific 1, Protease serine-like 1, PRSSL1protease, serine-like, 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Kallikrein 10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) rhKallikrein 3, rhKallikrein 5, and rhKallikrein 11 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
KLK10
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 10 Ala34-Asn276 Accession # O43240
Immunogen affinity purified
RUO
Primary
5655
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only