missing translation for 'onlineSavingsMsg'
Learn More

ISLR-2 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4526

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438774

  • £362.60 / 100µg

Estimated Shipment date
from Supplier 06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ISLR-2 Polyclonal antibody AF4526 Specifically detects ISLR-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

ISLR-2
LYOPH
Western Blot 0.1 ug/mL
Q6UXK2
immunoglobulin superfamily containing leucine-rich repeat 2, ISLR2, KIAA1465, leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein, LINX, UNQ1885/PRO4329
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ISLR-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human ISLR-2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ISLR2
Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590 Accession # Q6UXK2
Antigen Affinity-purified
RUO
Primary
57611
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only